Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CDS024714

Sigma-Aldrich

(1R)-3-Methyl-1-({(2S)-3-phenyl-2-[(2-pyrazinylcarbonyl)amino]propanoyl}amino)butylboronic acid

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H25BN4O4
Molecular Weight:
384.24
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

O=C(C1=CN=CC=N1)N[C@@H](CC2=CC=CC=C2)C(N[C@@H](CC(C)C)B(O)O)=O

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY, (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE, OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY, WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elisabet E Manasanch et al.
Nature reviews. Clinical oncology, 14(7), 417-433 (2017-01-25)
The ubiquitin proteasome pathway was discovered in the 1980s to be a central component of the cellular protein-degradation machinery with essential functions in homeostasis, which include preventing the accumulation of misfolded or deleterious proteins. Cancer cells produce proteins that promote
Noopur S Raje et al.
British journal of haematology, 176(5), 783-795 (2016-12-23)
In this double-blind, Phase 2 study, 220 patients with relapsed/refractory multiple myeloma were randomly assigned 1:1:1 to receive placebo (N = 72), tabalumab 100 mg (N = 74), or tabalumab 300 mg (N = 74), each in combination with dexamethasone 20 mg and subcutaneous bortezomib 1·3 mg/m
Craig T Wallington-Beddoe et al.
Oncotarget, 8(27), 43602-43616 (2017-05-04)
The proteasome inhibitor bortezomib has proven to be invaluable in the treatment of myeloma. By exploiting the inherent high immunoglobulin protein production of malignant plasma cells, bortezomib induces endoplasmic reticulum (ER) stress and the unfolded protein response (UPR), resulting in
Zhenrui Fu et al.
Anti-cancer drugs, 30(7), e0773-e0773 (2019-02-27)
Proteasome α5 subunit (PSMA5) is related to poor prognosis in various cancers. The first therapeutic proteasome inhibitor, bortezomib, induces apoptosis, suppressing cell growth in many tumor types. However, the effects of PSMA5 and bortezomib in prostate cancer (PCa) are still
Lisa P Deliu et al.
Biology open, 6(8), 1229-1234 (2017-06-24)
Translational control of gene expression is an important regulator of growth, homeostasis and aging in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service