Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CDS021691

Sigma-Aldrich

D-Glucosamine

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C6H13NO5
Molecular Weight:
179.17
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

description

AldrichCPR

form

solid

SMILES string

O[C@H]([C@@H](N)C([H])=O)[C@H](O)[C@H](O)CO

InChI

1S/C6H13NO5/c7-3(1-8)5(11)6(12)4(10)2-9/h1,3-6,9-12H,2,7H2/t3-,4+,5+,6+/m0/s1

InChI key

FZHXIRIBWMQPQF-SLPGGIOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY, (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE, OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY, WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicola Veronese et al.
Seminars in arthritis and rheumatism, 49(1), 9-19 (2019-02-05)
Type 2 diabetes mellitus (T2DM) and osteoarthritis (OA) are common diseases that frequently co-exist, along with overweight/obesity. While the mechanical impact of excess body weight on joints may explain lower limb OA, we sought to explore whether T2DM is linked
Anaïs Burgain et al.
PLoS pathogens, 15(12), e1007823-e1007823 (2019-12-07)
Inside the human host, the pathogenic yeast Candida albicans colonizes predominantly oxygen-poor niches such as the gastrointestinal and vaginal tracts, but also oxygen-rich environments such as cutaneous epithelial cells and oral mucosa. This suppleness requires an effective mechanism to reversibly
Cheng-Chau Chiu et al.
The journal of physical chemistry. A, 123(31), 6683-6700 (2019-07-12)
Motivated by the fundamental difference in the reactivity of hexoses and N-acetylhexosamines under collision-induced dissociation (CID) mass spectrometry conditions, we have investigated the CID of two hexosamines, glucosamine (GlcN) and galactosamine (GalN), experimentally and computationally. Both hexosamines undergo ring-opening and
Patrick B Wilson
Complementary therapies in medicine, 29, 152-157 (2016-12-04)
Sparse data are available to describe the prevalence of dietary supplementation among individuals with arthritis. Using a nationally representative sample, this investigation assessed the prevalence of dietary supplement use among Americans self-reporting arthritis. Data from 4606 participants of the 2011-2012
Shoko Kashiuchi et al.
Food & function, 10(8), 5008-5017 (2019-07-30)
Supplementation with cartilage constituents, such as glucosamine, chondroitin sulfate and collagen peptide, are believed to reduce pain associated with joint disorders, such as rheumatoid arthritis (RA). Here, we administered daily, 10 mg glucosamine or 100 mg chicken cartilage hydrolysate (CH)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service