Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CDS021592

Sigma-Aldrich

Apramycin

Synonym(s):

(2R,3R,4S,5S,6S)-5-Amino-2-((2R,3S,4R,4aR,6S,7R,8aS)-7-amino-6-((1R,2R,3S,4R,6S)-4,6-diamino-2,3-dihydroxycyclohexyloxy)-4-hydroxy-3-(me

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H41N5O11
Molecular Weight:
539.58
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

description

AldrichCPR

SMILES string

O[C@H]([C@@H]1NC)[C@](O[C@@H]2O[C@H]([C@@H](N)C3)[C@H](O)[C@@H](O)[C@@H]3N)([H])[C@@](C[C@H]2N)([H])O[C@@H]1O[C@H]([C@H](O)[C@H]4O)O[C@H](CO)[C@H]4N

InChI

1S/C21H41N5O11/c1-26-11-14(30)18-8(33-20(11)37-21-16(32)13(29)10(25)9(4-27)34-21)3-7(24)19(36-18)35-17-6(23)2-5(22)12(28)15(17)31/h5-21,26-32H,2-4,22-25H2,1H3/t5-,6+,7-,8+,9-,10-,11+,12+,13+,14-,15-,16-,17-,18+,19+,20-,21-/m1/s1

InChI key

XZNUGFQTQHRASN-XQENGBIVSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY, (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE, OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY, WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ammara Khalid et al.
ACS synthetic biology, 6(12), 2339-2349 (2017-10-12)
Terpenoids represent the largest class of natural products, some of which are resources for pharmaceuticals, fragrances, and fuels. Generally, mass production of valuable terpenoid compounds is hampered by their low production levels in organisms and difficulty of chemical synthesis. Therefore
Richard A Lewis et al.
PloS one, 14(4), e0200673-e0200673 (2019-04-11)
The AbsA1-AbsA2 two component signalling system of Streptomyces coelicolor has long been known to exert a powerful negative influence on the production of the antibiotics actinorhodin, undecylprodiginine and the Calcium-Dependent Antibiotic (CDA). Here we report the analysis of a ΔabsA2
Anna Rominski et al.
The Journal of antimicrobial chemotherapy, 72(8), 2191-2200 (2017-05-10)
Mycobacterium abscessus is innately resistant to a variety of drugs thereby limiting therapeutic options. Bacterial resistance to aminoglycosides (AGs) is conferred mainly by AG-modifying enzymes, which often have overlapping activities. Several putative AG-modifying enzymes are encoded in the genome of
Masaaki Ishikawa et al.
Scientific reports, 9(1), 2410-2410 (2019-02-23)
Spread of antimicrobial resistance and shortage of novel antibiotics have led to an urgent need for new antibacterials. Although aminoglycoside antibiotics (AGs) are very potent anti-infectives, their use is largely restricted due to serious side-effects, mainly nephrotoxicity and ototoxicity. We
Keita Amagai et al.
Scientific reports, 7(1), 3382-3382 (2017-06-15)
Telomestatin, a strong telomerase inhibitor with G-quadruplex stabilizing activity, is a potential therapeutic agent for treating cancers. Difficulties in isolating telomestatin from microbial cultures and in chemical synthesis are bottlenecks impeding the wider use. Therefore, improvement in telomestatin production and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service