Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CDS008176

Sigma-Aldrich

3-(isoxazol-4-yl)propanoic acid

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7NO3
Molecular Weight:
141.12
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

SMILES string

OC(=O)CCc1cnoc1

InChI

1S/C6H7NO3/c8-6(9)2-1-5-3-7-10-4-5/h3-4H,1-2H2,(H,8,9)

InChI key

YSXIJTDOCXJNHI-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shahid Ali Shah et al.
Molecular neurobiology, 51(3), 1278-1291 (2014-07-07)
Oxidative stress has been implicated in the pathophysiology of several neurodegenerative disorders. Numerous studies have reported that ethanol exposure produces reactive oxygen species (ROS), one of the best-known molecular mechanisms of ethanol neurotoxicity. We recently reported gamma-aminobutyric acid B1 receptor
Da-Bing Li et al.
Scientific reports, 9(1), 6029-6029 (2019-04-17)
It has been well established that the cerebellum and its associated circuitry constitute the essential neuronal system for both delay and trace classical eyeblink conditioning (DEC and TEC). However, whether the cerebellum is sufficient to independently modulate the DEC, and
Dool-Ri Oh et al.
The American journal of Chinese medicine, 49(1), 69-94 (2020-12-30)
Vaccinium bracteatum Thunb. Leaves (VBL) are a component of traditional herbal medicines. However, molecular mechanisms of VBL in stress-related memory impairment are still unclear. This study aimed to investigate the spatial memory improvement effects of VBL in an animal model
Alexandra Colón-Rodríguez et al.
Neurotoxicology, 81, 116-126 (2020-09-30)
α motor neurons (MNs) are a target of the environmental neurotoxicant methylmercury (MeHg), accumulating MeHg and subsequently degenerating. In mouse spinal cord MN cultures, MeHg increased intracellular Ca2+ [Ca2+]i; the AMPA receptor (AMPAR) antagonist CNQX delayed the increase in [Ca2+]i

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service