Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CDS006817

Sigma-Aldrich

Fmoc-His-OH

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H19N3O4
Molecular Weight:
377.39
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

functional group

Fmoc

SMILES string

OC(=O)[C@H](Cc1c[nH]cn1)NC(=O)OCC2c3ccccc3-c4ccccc24

InChI

1S/C21H19N3O4/c25-20(26)19(9-13-10-22-12-23-13)24-21(27)28-11-18-16-7-3-1-5-14(16)15-6-2-4-8-17(15)18/h1-8,10,12,18-19H,9,11H2,(H,22,23)(H,24,27)(H,25,26)/t19-/m0/s1

InChI key

SIRPVCUJLVXZPW-IBGZPJMESA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Fabian Schumacher et al.
Journal of lipid research, 58(8), 1648-1660 (2017-06-08)
Sphingosine 1-phosphate (S1P), a bioactive lipid involved in various physiological processes such as cell proliferation and apoptosis, can be irreversibly cleaved by S1P lyase, yielding phosphoethanolamine and (2
Mohammed Dwidar et al.
Journal of the American Chemical Society, 141(28), 11103-11114 (2019-06-27)
Artificial cells that encapsulate DNA-programmable protein expression machinery are emerging as an attractive platform for studying fundamental cellular properties and applications in synthetic biology. However, interfacing these artificial cells with the complex and dynamic chemical environment remains a major and
Alex W Wilkinson et al.
Nature, 565(7739), 372-376 (2019-01-11)
For more than 50 years, the methylation of mammalian actin at histidine 73 has been known to occur1. Despite the pervasiveness of His73 methylation, which we find is conserved in several model animals and plants, its function remains unclear and the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service