Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CDS005354

Sigma-Aldrich

Fmoc-(S)-3-amino-3-(6-methoxy-3-pyridyl)-propionic acid

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H22N2O5
Molecular Weight:
418.44
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

functional group

Fmoc

SMILES string

COc1ccc(cn1)[C@H](CC(O)=O)NC(=O)OCC2c3ccccc3-c4ccccc24

InChI

1S/C24H22N2O5/c1-30-22-11-10-15(13-25-22)21(12-23(27)28)26-24(29)31-14-20-18-8-4-2-6-16(18)17-7-3-5-9-19(17)20/h2-11,13,20-21H,12,14H2,1H3,(H,26,29)(H,27,28)/t21-/m0/s1

InChI key

XSOWGPXMQMVHDC-NRFANRHFSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service