Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CDS004770

Sigma-Aldrich

4-pyridylethylmercaptan

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9NS
CAS Number:
Molecular Weight:
139.22
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

liquid

SMILES string

SCCc1ccncc1

InChI

1S/C7H9NS/c9-6-3-7-1-4-8-5-2-7/h1-2,4-5,9H,3,6H2

InChI key

VBAOEVKQBLGWTH-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huey Wen Ooi et al.
Biomacromolecules, 17(1), 280-290 (2015-12-03)
We report the development of thermoresponsive 4-mercaptoethylpyridine (MEP)-based chromatographic microsphere based resins for antibody separation that show switchable release abilities by adsorbing immunoglobulins at 40 °C and releasing the proteins at 5 °C. The thermoswitchable release properties were introduced to
Martin Burkhardt et al.
Vaccine, 37(38), 5762-5769 (2018-09-29)
Efforts to develop a vaccine for the elimination of malaria include the use of carrier proteins to assemble monomeric antigens into nanoparticles to maximize immunogenicity. Recombinant ExoProtein A (EPA) is a detoxified form of Pseudomonas aeruginosa Exotoxin A which has
K van der Maaden et al.
Langmuir : the ACS journal of surfaces and colloids, 35(22), 7121-7128 (2019-05-03)
For pharmaceutical, biological, and biomedical applications, the functionalization of gold surfaces with pH-sensitive groups has great potential. The aim of this work was to modify gold surfaces with pH-sensitive groups and to determine the p Ka of the modified gold
Elodie Dumont et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 233, 118180-118180 (2020-03-13)
Nowadays, the use of functionalised surface-enhanced Raman scattering (SERS) substrates has become common. These surface modifying agents notably act as Raman reporters, as sensors of biological processes (pH, redox probes) or to increase the sensitivity and/or the specificity of SERS

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service