Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CDS004670

Sigma-Aldrich

1,2-diaminonaphthalene

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10N2
Molecular Weight:
158.20
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

Nc1ccc2ccccc2c1N

InChI

1S/C10H10N2/c11-9-6-5-7-3-1-2-4-8(7)10(9)12/h1-6H,11-12H2

InChI key

NTNWKDHZTDQSST-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

pictograms

Health hazardExclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wojciech Michno et al.
Biochimica et biophysica acta. Proteins and proteomics, 1867(5), 458-467 (2018-10-03)
While the molecular mechanisms underlying Alzheimer's disease (AD) remain largely unknown, abnormal accumulation and deposition of beta amyloid (Aβ) peptides into plaques has been proposed as a critical pathological process driving disease progression. Over the last years, neuronal lipid species
Mária Trundová et al.
International journal of biological macromolecules, 114, 776-787 (2018-03-28)
The Gram-negative bacterium Legionella pneumophila is one of the known opportunistic human pathogens with a gene coding for a zinc-dependent S1-P1 type nuclease. Bacterial zinc-dependent 3'-nucleases/nucleotidases are little characterized and not fully understood, including L. pneumophila nuclease 1 (Lpn1), in
Kosta Theodorou et al.
Scientific reports, 7(1), 11670-11670 (2017-09-17)
Although A Disintegrin And Metalloproteinase 8 (ADAM8) is not crucial for tissue development and homeostasis, it has been implicated in various inflammatory diseases by regulating processes like immune cell recruitment and activation. ADAM8 expression has been associated with human atherosclerosis
R S Soumya et al.
International journal of biological macromolecules, 109, 740-747 (2017-11-21)
The field of nanotechnology has overgrown over the past few years and has even ventured into the field of medicine. The aim of the present study is to develop a novel allicin functionalized locust bean gum nanoparticle using the nanoprecipitation
Raiza Raianne Luz Rodrigues et al.
International immunopharmacology, 90, 107124-107124 (2020-11-11)
Leishmaniasis is caused by several protozoan species of Leishmania, and being endemically present in 98 countries around the world, it is also a severe public-health problem. The available antileishmanial drugs are toxic and yet present risks of recurrent infection. Efforts

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service