Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CDS004506

Sigma-Aldrich

N-hydroxyacetamide

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H6N2O
CAS Number:
Molecular Weight:
74.08
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

NC(CO)=O

InChI

1S/C2H5NO2/c3-2(5)1-4/h4H,1H2,(H2,3,5)

InChI key

TZGPACAKMCUCKX-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naoki Nakagawa et al.
Kidney international, 87(6), 1125-1140 (2015-02-05)
MicroRNAs, activated by the enzyme Dicer1, control post-transcriptional gene expression. Dicer1 has important roles in the epithelium during nephrogenesis, but its function in stromal cells during kidney development is unknown. To study this, we inactivated Dicer1 in renal stromal cells.
Emily P Zeitler et al.
JACC. Heart failure, 3(2), 159-167 (2014-12-30)
The purpose of this study was to assess the benefit of primary prevention implantable cardioverter defibrillators (ICDs) in women. Clinical trials of primary prevention ICDs enrolled a limited number of women. Using a propensity score method, we matched 490 women ≥65
L M Verweij et al.
Netherlands heart journal : monthly journal of the Netherlands Society of Cardiology and the Netherlands Heart Foundation, 23(9), 420-427 (2015-05-30)
To assess the comparability of five performance indicator scores for treatment delay among patients diagnosed with ST-segment elevation myocardial infarction (STEMI) undergoing primary percutaneous coronary intervention in relation to the quality of the underlying data. Secondary analyses were performed on
Yunzhou Dong et al.
The Journal of clinical investigation, 125(12), 4349-4364 (2015-11-17)
Tumor angiogenesis is critical for cancer progression. In multiple murine models, endothelium-specific epsin deficiency abrogates tumor progression by shifting the balance of VEGFR2 signaling toward uncontrolled tumor angiogenesis, resulting in dysfunctional tumor vasculature. Here, we designed a tumor endothelium-targeting chimeric
Gary S Ma et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(20), E2602-E2610 (2015-05-01)
In eukaryotes, receptor tyrosine kinases (RTKs) and trimeric G proteins are two major signaling hubs. Signal transduction via trimeric G proteins has long been believed to be triggered exclusively by G protein-coupled receptors (GPCRs). This paradigm has recently been challenged

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service