Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CDS003223

Sigma-Aldrich

4-Piperidone

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO
Molecular Weight:
99.13
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

O=C1CCNCC1

InChI

1S/C5H9NO/c7-5-1-3-6-4-2-5/h6H,1-4H2

Inchi Key

VRJHQPZVIGNGMX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yangjian Zhou et al.
Chemosphere, 235, 900-907 (2019-07-14)
Effluent organic matter (EfOM), which is composed of background natural organic matter (NOM), soluble microbial degradation products, and trace amounts of organic pollutants, can play an important role in the photodegradation of emerging pollutants in the effluent. In this study
Djeda Belharazem et al.
Oncotarget, 8(52), 89580-89594 (2017-11-23)
The anti-apoptotic cellular FLICE-like inhibitory protein cFLIP plays a pivotal role in normal tissues homoeostasis and the development of many tumors, but its role in normal thymus (NT), thymomas and thymic carcinomas (TC) is largely unknown. Expression, regulation and function
Brian P Mayer et al.
Talanta, 186, 645-654 (2018-05-23)
Chemical attribution of the origin of an illegal drug is a key component of forensic efforts aimed at combating illicit and clandestine manufacture of drugs and pharmaceuticals. The results of these studies yield detailed information on synthesis byproducts, reagents, and
T S Mayadevi et al.
Membranes, 10(11) (2020-11-11)
A series of poly(meta/para-terphenylene-methyl piperidinium)-based anion exchange membranes devoid of benzylic sites or aryl ether bonds, that are vulnerable to degradation by hydroxide ions, are synthesized and investigated for their application as novel anion exchange membranes. The copolymers are composed
Babu Balaji et al.
European journal of medicinal chemistry, 92, 332-341 (2015-01-13)
Oxovanadium(IV) complexes [VO(Fc-tpy)(acac)](ClO4) (1), [VO(Fc-tpy)(nap-acac)](ClO4) (2), [VO(Fc-tpy)(py-acac)](ClO4) (3) and [VO(Ph-tpy)(py-acac)](ClO4) (4) of 4'-ferrocenyl-2,2':6',2"-terpyridine (Fc-tpy) and 4'-phenyl-2,2':6',2"-terpyridine (Ph-tpy) having monoanionic acetylacetonate (acac), naphthylacetylacetonate (nap-acac) or pyrenylacetylacetonate (py-acac) ligand were prepared, characterized and their photocytotoxicity in visible light studied. The ferrocenyl complexes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service