Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CDS002826

Sigma-Aldrich

3-Nitro-1H-pyrazole

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H3N3O2
Molecular Weight:
113.07
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

[O-][N+](=O)c1cc[nH]n1

InChI

1S/C3H3N3O2/c7-6(8)3-1-2-4-5-3/h1-2H,(H,4,5)

InChI key

MZRUFMBFIKGOAL-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

320.0 °F - closed cup

flash_point_c

160 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark Zanin et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(42), 11217-11222 (2017-09-07)
North American wild birds are an important reservoir of influenza A viruses, yet the potential of viruses in this reservoir to transmit and cause disease in mammals is not well understood. Our surveillance of avian influenza viruses (AIVs) at Delaware
Shinobu Masuda et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 67(8), 563-574 (2019-06-12)
Breast cancer (BC) is a heterogeneous disease with evolving genetic alterations and expressions of receptor proteins. Intratumoral heterogeneity (ITH) is considered to be a resistance factor in response to targeted therapies. The current single-slide, single-marker immunohistochemistry techniques cannot accurately assess
Ubaradka G Sathyanarayana et al.
Applied immunohistochemistry & molecular morphology : AIMM, 23(5), 364-373 (2014-09-30)
Prostate biopsy is the key clinical specimen for disease diagnosis. However, various conditions used during biopsy processing for histologic analysis may affect the performance of diagnostic tests, such as hematoxylin and eosin (H&E) staining, immunohistochemistry (IHC), or in situ hybridization

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service