Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CDS000473

Sigma-Aldrich

Coriphosphine O

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18ClN3
Molecular Weight:
287.79
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

SMILES string

Cl.CN(C)c1ccc2cc3cc(C)c(N)cc3nc2c1

InChI

1S/C16H17N3.ClH/c1-10-6-12-7-11-4-5-13(19(2)3)8-15(11)18-16(12)9-14(10)17;/h4-9H,17H2,1-3H3;1H

InChI key

JRMDFAKCPRMZKA-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prajakta S Jogdand et al.
Malaria journal, 11, 235-235 (2012-07-24)
Functional in vitro assays could provide insights into the efficacy of malaria vaccine candidates. For estimating the anti-parasite effect induced by a vaccine candidate, an accurate determination of live parasite count is an essential component of most in vitro bioassays.
Takahiro Tougan et al.
Scientific reports, 10(1), 15605-15605 (2020-09-26)
Iron deficiency is the most common and widespread nutritional disorder worldwide. The automated haematology analyser XN-30 (Sysmex, Kobe, Japan) was developed to detect malaria-infected red blood cells (RBCs) in human blood samples using flow cytometry. The optical system of the
Viviana Martins et al.
Plant physiology and biochemistry : PPB, 150, 49-55 (2020-03-03)
Having a central role in cell wall pectin cross-linking, calcium has been increasingly used as supplement to promote fruit firmness and extended shelf-life. However, the molecular rearrangements associated to increased fruit robustness are still a matter of debate. In this
Amanda A Cardoso et al.
The New phytologist, 227(6), 1804-1817 (2020-05-10)
Lycophytes are the earliest diverging extant lineage of vascular plants, sister to all other vascular plants. Given that most species are adapted to ever-wet environments, it has been hypothesized that lycophytes, and by extension the common ancestor of all vascular

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service