Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CBR01086

Sigma-Aldrich

1-(4-Methyl-1,3-thiazol-5-yl)ethanone

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7NOS
CAS Number:
Molecular Weight:
141.19
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

O=C(C)C1=C(C)N=CS1

InChI

1S/C6H7NOS/c1-4-6(5(2)8)9-3-7-4/h3H,1-2H3

InChI key

MBLHLRLBSIWLMG-UHFFFAOYSA-N

Analysis Note

may contain up to 0.5 eq water

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Legal Information

Product of ChemBridge Corp.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S E Tsuei et al.
Journal of chromatography, 182(1), 55-62 (1980-04-11)
A simple and rapid gas chromatographic method for the quantitation of chlormethiazole and two of its pharmacologically active metabolites, 5-acetyl-4-methylthiazole and 5-(1-hydroxyethyl)-4-methylthiazole, in plasma and blood is described. The total analysis time is less than 20 min for a single
A R Green et al.
British journal of pharmacology, 129(1), 95-100 (2000-02-29)
A single dose of clomethiazole (600 micromol kg(-1) i.p.) has previously been shown to be neuroprotective in the gerbil model of global ischaemia. In gerbils, clomethiazole (600 micromol kg(-1)) injection produced a rapid appearance (peak within 5 min) of drug

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service