Key Documents
CAD7971
Micro-emulsifying needle
without reinforcing bar, gauge 22, L 2 7/8 in.
About This Item
UNSPSC Code:
42142546
NACRES:
NB.03
packaging
pack of 4 ea
manufacturer/tradename
Cadence Science 7971
L
2 7/8 in.
gauge
22
General description
Widely used for allergens and the mixing of adjuvants and antigens, oil based liquids or any two liquid media differing in viscosity. By locking two syringes on to the double hub needle or double hub adapter, two liquids such as a mineral oil and an emulsifying agent may be readily pumped between the two syringes to produce a uniform mixture.
Indicated lengths are the overall length. Researchers and technicians utilize Micro-Mate® 5 mL and 10 mL Luer lock tip glass syringes for micro-emulsifying procedures. The outstanding quality of the glass and the special secure Luer locking tip provide quality and reliability.
Legal Information
Micro-Mate is a registered trademark of Cadence Science
Choose from one of the most recent versions:
Certificates of Analysis (COA)