Key Documents
CAD4040
Standard point hypodermic needles, 304 stainless steel
gauge 23, L 2 in., nickel plated hub
Select a Size
12 EA
$440.00
$440.00
Available to ship onApril 29, 2025Details
Select a Size
Change View
12 EA
$440.00
About This Item
UNSPSC Code:
42142523
NACRES:
NB.03
$440.00
Available to ship onApril 29, 2025Details
material
304 stainless steel needle
nickel plated hub
packaging
pack of 12 ea
manufacturer/tradename
Cadence Science 4040
L
2 in.
gauge
23
General description
Hypodermic needles are precision made and designed to ensure strict adherence with U.S. Government specification GG-N-196. Points are machine ground to achieve uniform sharpness. Precision grinding, micro-bead jet blasting, and ultrasonic cleaning assure freedom from burrs and debris. All needle cannula are fabricated of type 304 stainless steel tubing which must pass rigorous quality control prior to acceptance and use.
Standard nickle plated Luer hubs comply with Luer taper specifications ISO 594. This assures an interchangeable fit mating with Luer slips and Luer locks. Contoured hub cavity facilitates cleaning.
Choose from one of the most recent versions:
Certificates of Analysis (COA)