Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

C72404

Sigma-Aldrich

2-Chlorothioxanthen-9-one

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H7ClOS
CAS Number:
Molecular Weight:
246.71
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

assay

98%

form

powder

mp

153-154 °C (lit.)

SMILES string

Clc1ccc2Sc3ccccc3C(=O)c2c1

InChI

1S/C13H7ClOS/c14-8-5-6-12-10(7-8)13(15)9-3-1-2-4-11(9)16-12/h1-7H

InChI key

ZCDADJXRUCOCJE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Sol. 2

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 3

flash_point_f

195.8 °F - closed cup

flash_point_c

91 °C - closed cup

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J E Pettersen et al.
Journal of pharmaceutical sciences, 70(7), 812-813 (1981-07-01)
In gastric aspirate from a case of severe chlorprothixene poisoning, large amounts (approximately 30% of the chlorprothixene) of a previously unrecognized compound were found and identified tentatively as 2-chlorothioxanthen-9-one by combined GLC-low-resolution mass spectrometry and high-resolution mass spectrometry. The identity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service