Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C59408

Sigma-Aldrich

4-Chloro-3-nitrobenzenesulfonyl chloride

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(NO2)SO2Cl
CAS Number:
Molecular Weight:
256.06
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

96%

form

powder

mp

59-60 °C (lit.)

SMILES string

[O-][N+](=O)c1cc(ccc1Cl)S(Cl)(=O)=O

InChI

1S/C6H3Cl2NO4S/c7-5-2-1-4(14(8,12)13)3-6(5)9(10)11/h1-3H

InChI key

SEWNAJIUKSTYOP-UHFFFAOYSA-N

Application

4-Chloro-3-nitrobenzenesulfonyl chloride can be used:
  • In the preparation of functionalized 1H-indenes via copper-catalyzed arylative cyclization.[1]
  • As a precursor in the synthesis of N-arylsulfonyl-3-acylindole arylcarbonyl hydrazone derivatives that show potential nematicidal activity.[2]
  • As a key starting material in the synthesis of sulfonamide analogs of ferrostatin-1 (fer-1), which possess good ferroptosis inhibition property.[3]
  • In the synthesis of 4-chloro-3-nitrobenzene thiol.[4]

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel ferroptosis inhibitors with improved potency and ADME properties.
Hofmans, Sam et al.
Journal of Medicinal Chemistry, 59(5), 2041-2053 (2016)
Synthesis and quantitative structure-activity relationship (QSAR) study of novel N-arylsulfonyl-3-acylindole arylcarbonyl hydrazone derivatives as nematicidal agents.
Che, Zhiping et al.
Journal of Agricultural and Food Chemistry, 61(24), 5696-5705 (2013)
A simple, fast and chemoselective method for the preparation of arylthiols.
Bellale, Eknath V et al.
Synthesis, 2009(19), 3211-3213 (2009)
Xiaoming Zeng et al.
Journal of the American Chemical Society, 133(44), 17638-17640 (2011-10-13)
A variety of polysubstituted 1H-indenes can be prepared through the copper-catalyzed arylative cyclization of simple arylalkynes with commercially available aromatic sulfonyl chlorides that function as an aryl group donor. The reaction tolerates a broad range of functional groups, including bromide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service