Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C57200

Sigma-Aldrich

4-Chloro-α-methylstyrene

98%, contains 100 ppm tert-butylcatechol as stabilizer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(CH3)C6H4Cl
CAS Number:
Molecular Weight:
152.62
Beilstein/REAXYS Number:
1855073
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

assay

98%

contains

100 ppm tert-butylcatechol as stabilizer

refractive index

n20/D 1.555 (lit.)

density

1.065 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=C)c1ccc(Cl)cc1

InChI

1S/C9H9Cl/c1-7(2)8-3-5-9(10)6-4-8/h3-6H,1H2,2H3

InChI key

WQDGTJOEMPEHHL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

163.4 °F - closed cup

flash_point_c

73 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service