Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C3103

Sigma-Aldrich

Carbazole

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9N
CAS Number:
Molecular Weight:
167.21
Beilstein/REAXYS Number:
3956
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

vapor pressure

400 mmHg ( 323 °C)

assay

96%

form

powder

bp

355 °C (lit.)

mp

243-246 °C (lit.)

SMILES string

c1ccc2c(c1)[nH]c3ccccc23

InChI

1S/C12H9N/c1-3-7-11-9(5-1)10-6-2-4-8-12(10)13-11/h1-8,13H

InChI key

UJOBWOGCFQCDNV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guodong Ji et al.
Journal of hazardous materials, 225-226, 1-7 (2012-05-23)
We examined the effects of power and treatment time on the ultrasonically enhanced ozonation of carbazole dissolved in APG(1214) surfactant solutions, including an analysis of the mechanism of OH radical formation, the zeta potential of the colloidal suspension, the influence
Yuji Ashikawa et al.
BMC structural biology, 12, 15-15 (2012-06-26)
Dihydroxylation of tandemly linked aromatic carbons in a cis-configuration, catalyzed by multicomponent oxygenase systems known as Rieske nonheme iron oxygenase systems (ROs), often constitute the initial step of aerobic degradation pathways for various aromatic compounds. Because such RO reactions inherently
Lena Arnold et al.
Chemical communications (Cambridge, England), 48(77), 9640-9642 (2012-08-22)
Carbazole-containing porphyrinoid was synthesised for the first time via Suzuki-Miyaura cross-coupling reaction. Oxidation with MnO(2) yielded its porphyrin-state featuring macrocyclic aromaticity in exchange for the loss of the resonance energies of the benzene rings.
Jae-Min Suk et al.
Organic letters, 14(19), 5018-5021 (2012-09-18)
Chiral organic anions such as camphorsulfonates and cAMP give rise to the preferential formation of a one-handed helix of an indolocarbazole foldamer, thus inducing characteristic circular dichroic (CD) signals. Moreover, the on and off switching of the chiroptical signal can
Jong-Kwan Bin et al.
Advanced materials (Deerfield Beach, Fla.), 24(21), 2911-2915 (2012-05-03)
A new carbazole derivative with two carbazole moieties on the C3 and C6 positions of carbazole and triphenylsilane directly linked to the N of carbazole is successfully used as a highly efficient blue phosphorescent host in an organic light-emitting diode

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service