Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C101109

Sigma-Aldrich

1,3-Cyclohexanediol, mixture of cis and trans

98%

Synonym(s):

Hexahydroresorcinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H10(OH)2
CAS Number:
Molecular Weight:
116.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

solid

bp

246-247 °C (lit.)

SMILES string

OC1CCCC(O)C1

InChI

1S/C6H12O2/c7-5-2-1-3-6(8)4-5/h5-8H,1-4H2

InChI key

RLMGYIOTPQVQJR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael J Radcliffe et al.
BMJ (Clinical research ed.), 327(7409), 251-254 (2003-08-05)
To assess the efficacy of enzyme potentiated desensitisation in the treatment of severe summer hay fever poorly controlled by pharmacotherapy. Double blind randomised placebo controlled parallel group study. Hospital in Hampshire. 183 participants aged between 18 and 64 with a
G A Mills et al.
Clinical chemistry, 36(6), 870-874 (1990-06-01)
Using gas chromatography-mass spectrometry, we investigated the urinary excretion of organic acids of 278 newborn babies in a special care unit to obtain reference data and monitor metabolism. In 101 of 584 urine samples analyzed, we found isomers of cyclohexanediol.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service