Key Documents
BR987402
BRAND® hydrometer jar
size 250 mL
Synonym(s):
Accessories, Hydrometer jar, Hydrometers
About This Item
UNSPSC Code:
41113024
material
glass
packaging
pack of 1 ea
manufacturer/tradename
BRAND 987402
size
250 mL
General description
Ungraduated with hexagonal base and spout.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)