Key Documents
BR980360
BRAND® general purpose alcoholometer Gay-Lussac+Cartier
with thermometer
Synonym(s):
Alcoholometers, Gay-Lussac+Cartier pattern, Hydrometers
About This Item
UNSPSC Code:
41121500
description
range 0-100 vol.%/10-45 °Cartier
packaging
pack of 1 ea
manufacturer/tradename
BRAND 980360
L
330 mm
General description
Subdivisions: 1 vol./0.5 °Cartier, reference temperature 15 °C, range 0-100 vol.%/10-45 °Cartier.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)