Key Documents
BR969612
BRAND® general purpose hydrometer with thermometer
1.000 to 1.200 g/cm3
Synonym(s):
Hydrometers
About This Item
UNSPSC Code:
41113024
description
0-30 °C subdivision 1 °C
packaging
pack of 1 ea
manufacturer/tradename
BRAND 969612
L
330 mm
General description
Subdivisions 0.002 g/cm3, reference temperature 20 °C. With temperature. Thermometer range from 0-30/40 °C (subdivision 1 °C). filling: petroleum, colored blue.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)