Key Documents
BR87812
BRAND® PP beaker with spout, low form
volume 250 mL
Synonym(s):
Beakers, Plastics
About This Item
UNSPSC Code:
41121803
NACRES:
NB.43
material
polypropylene
description
ISO 7056
feature
graduations
beaded rim and spout
manufacturer/tradename
BRAND 87812
volume
250 mL
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)