Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BR781642

BRAND® microplate BRANDplates®, pureGrade

size 1536 wells, size 10 μL, flat bottom, black

Synonym(s):

BRANDplates®, Microplates, medium binding, pureGrade

Sign Into View Organizational & Contract Pricing

Select a Size

50 EA
$1,280.00

$1,280.00


Check Cart for Availability


Select a Size

Change View
50 EA
$1,280.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
material:
black wells
flat bottom
polystyrene
size:
10 μL
1536 wells

$1,280.00


Check Cart for Availability

material

black wells
flat bottom
polystyrene

sterility

non-sterile

feature

lid: no

packaging

pack of 50 ea

manufacturer/tradename

BRAND 781642

size

10 μL
1536 wells

working volume

10 μL

color

black

suitability

suitable for (bacteriological assays)

binding type

medium binding surface

Looking for similar products? Visit Product Comparison Guide

General description

The BRAND® Microplate BRANDplates® PureGrade features a medium-binding, untreated surface, making it an ideal choice for a variety of applications. This non-sterile microplate serves as the standard for use in homogeneous assays, screenings, and sample storage. It is non-cytotoxic, making it safe for use in cell-based assays. With BIO-CERT® CERTIFIED QUALITY, this microplate is guaranteed to be free from human DNA, RNase, DNase, and pyrogens. This versatile microplate is a dependable option for researchers looking for consistent performance across numerous laboratory applications.

Features and Benefits

  • Optimal surfaces for optimal results
  • Reliable quality from the cleanroom
  • Tested by independent laboratories

Legal Information

BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service