BR781391
BRAND® sealing film for microplates
self-adhesive, suitable for (real-time PCR), polyester
Synonym(s):
Accessories, PCR, Sealing film
About This Item
material
polyester
polyester film
sterility
non-sterile
feature
piercable
self-adhesive
packaging
pack of 100 ea
manufacturer/tradename
BRAND 781391
technique(s)
PCR: suitable
suitability
suitable for (real-time PCR)
Looking for similar products? Visit Product Comparison Guide
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service