BR781347
BRAND® 384-well PCR plate
full skirt, cut corner A12, clear
Synonym(s):
PCR, PCR Plates, Plates
About This Item
Recommended Products
material
clear
polypropylene
description
D-Nase/R-Nase, DNA-free, suitable for qPCR
sterility
non-sterile
feature
skirt
full skirt, cut corner A12
packaging
pack of 50 ea
manufacturer/tradename
BRAND 781347
technique(s)
PCR: suitable
size
384 well
working volume
30 μm
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service