Key Documents
BR780400
BRAND® disposable microcentrifuge tube
with safety lid, capacity 1.5 mL, sterile
Synonym(s):
Microcentrifuge Tubes, Tubes
About This Item
UNSPSC Code:
41121703
NACRES:
NB.11
material
polypropylene
sterility
sterile
feature
with safety lid
packaging
pack of 450
manufacturer/tradename
BRAND 780400
parameter
20000 × g max. RCF
capacity
1.5 mL
suitability
suitable for (free of endotoxins, DNA, Rnase and ATP)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)