BR778713
BRAND® centrifuge tube, beaded rim
volume 15 mL, conical bottom, graduated
Synonym(s):
Centrifuge tubes, Tubes
All Photos(1)
About This Item
UNSPSC Code:
41121703
material
borosilicate glass
conical bottom
feature
RCF 3,500 × g
graduated
packaging
pack of 10
manufacturer/tradename
BRAND 778713
volume
15 mL
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)