Key Documents
BR759701
BRAND® stand for disposal bags
includes pack of 50 bags, PA
Synonym(s):
Accessories, Disposable Bags, Stands
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
material
epoxy-coated (steel wire, with rubber feet)
feature
includes pack of 50 bags, PA
packaging
pack of 50 bags
manufacturer/tradename
BRAND 759701
General description
Bags must be open during sterilization. Autoclavable (134°C) and hot-air sterilization at 180°C.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)