Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BR732654

BRAND® Robotic Liquid Handling Station Filter Tips

non-sterile, colorless, DNA free, RNase free, endotoxin free, pkg of 10 × 96 pieces, volume 40-1000 μL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121601

sterility

non-sterile

feature

DNA free
RNase free
endotoxin free

packaging

pkg of 10 × 96 pieces

manufacturer/tradename

BRAND

volume

40-1000 μL

color

colorless

Looking for similar products? Visit Product Comparison Guide

General description

All BRAND robotic tips are produced using the most advanced cleanroom techniques to ensure consistently high quality for automation processes. The tips are available, non-sterile or BIO-CERT sterile, and non-filter or filter. They are manufactured in an environmentally friendly manner using only high-purity polypropylene, free from oleamide and diHEMDA – additives which can interfere with biological tests. BRAND robotic tips are specially designed and inspected for optimal tip fit with BRAND liquid end pipetting modules.

Product Features:

Manufactured without lubricants.
Cadmium-free pigments.
Graduation for quick volume check.
All rack-packed tips are free from DNA(<40fg), RNase(<8.6fg), ATP(<1fg) and endotoxins(<1fg).
Autoclavable at 121°C (2bar), acc. DIN EN 285.

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service