Key Documents
BR709101
BRAND® disposable BLAUBRAND® micropipettes, intraEND
capacity 1 μL
Synonym(s):
Micropipettes, Pipettes, intraEND
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
feature
DE-M marking, To Contain
packaging
pack of 1000 ea (4 boxes of 250)
manufacturer/tradename
BRAND 709101
L
32 mm
capacity
1 μL
General description
DE-M marking certified for volumes greater than 5 μl. Volume defined by end-to-end filling. Calibrated to contain (TC, In).
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)