Key Documents
BR708462
BRAND® EASYCAL testing tubes for pipettes < 50 μL
Synonym(s):
Calibration Software, EASYCAL
Select a Size
250 EA
$53.10
$53.10
Check Cart for Availability
Select a Size
Change View
250 EA
$53.10
About This Item
UNSPSC Code:
41121700
NACRES:
NB.01
$53.10
Check Cart for Availability
packaging
pack of 250 ea
manufacturer/tradename
BRAND 708462
General description
Avoid time-consuming evaporation traps or an expensive dual-pan balance! Pipettes < 50 μL can be surprisingly easy to calibrate using EASYCAL testing tubes (available as an accessory) or using the new micro-weighing container.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
EASYCAL is a trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)