Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BR704793

BRAND® TRANSFERPETTE® S starter kits

standard, volumes 0.5-10 μL, 10-100 μL and 100-1000 μL

Synonym(s):

Air displacement pipettes, Air interface pipettes, Microliter pipettes, Multichannel pipettes, Pipettes, Piston-operated pipettes, Single channel pipettes, Transferpette® S

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

feature

electronic: no
single-channel

packaging

pack of 3 ea

manufacturer/tradename

BRAND 704793

volume

0.5-1000 μL
10-100 μL
100-1000 μL

General description

The Transferpette S microliter pipettes are manual single and multi-channel pipettes for productive and precise pipetting in the laboratory. They offer high quality and many features that make work easier and more efficient, for example: Genuine one-hand operation, short stroke, low operating forces, volume adjustment protection and an easy-to-read volume display. They are easy to maintain and can be adjusted without tools using Easy Calibration technology. The single-channel Transferpette S pipettes cover the volume range from 0.1 ul to 10 ml. The Transferpette S -8 and -12 multichannel pipettes are available in volumes from 0.5 ul to 300 ul.
contains - 3 Transferpette® S adjustable pipettes (0.5-10 μL, 10-100 μL and 100-1000 μL), 3 x TipBox (filled), 3 rack mounts.

Features and Benefits

  • Efficient: one-hand operation
  • Flexible: ergonomic design for all hand sizes
  • Reliable: volume-change protection prevents accidental adjustments

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG
Transferpette is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

This video explains how to adjust the volume on the Transferpette before being ready to pipette.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service