Key Documents
BR704268
BRAND® support rod connector for remote dispensing Dispensette®
Synonym(s):
Accessories, Bottle-top Dispenser, Dispenser, Dispensette®
Select a Size
1 EA
$109.00
$109.00
Check Cart for Availability
Select a Size
Change View
1 EA
$109.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.43
$109.00
Check Cart for Availability
material
stainless steel
packaging
pack of 1 ea
manufacturer/tradename
BRAND 704268
Related Categories
General description
Dispense accurate volumes directly from drums and bulk refills.The Dispensette® can be mounted on a wall, a ring stand or on lab furniture.A filter in the drum adapter minimizes risk of contaminating highpurity reagents when air is drawn in for pressure compensation after dispensing.A quick-release connector with integrated valves allows quick changing of the bulk container.The remote dispensing system allows storage of the drum up to 10 meters (30 feet) away from the Dispensette®.The max. delivery height is approximately 1.2 m.Operating Exclusions: Never use the remote dispensing system: 1. with recirculation valve. It has to be removed before use! 2. for pressurized vessels 3. for liquids attacking borosilicate glass, Al2O3-ceramic, PFA, ETFE, FEP or PTFE 4. for Peroxide (due to catalytic reaction) 5. for carbon disulfide (CS2), due to risk of explosion!
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)