Key Documents
BR702406
BRAND® PD-Tip
maximum volume 1.0 mL, non-sterile
Synonym(s):
Accessories, Dispenser, HandyStep®, PD-Tips, Positive displacement principle, Repetitive Pipettes, Tips
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
material
colorless
polypropylene
description
includes adapter: No
material cylinder (PP) piston (PE-HD)
sterility
non-sterile
packaging
pack of 100 ea
manufacturer/tradename
BRAND 702406
maximum volume
1.0 mL
General description
cylinder (PP) piston (PE-HD)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
HandyStep is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)