Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BR701501

BRAND® PARAFILM® M sealing film

L × W 15 m × 500 mm

Synonym(s):

PARAFILM® M, Sealing films

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000

form

film

manufacturer/tradename

BRAND 701501

L × W

15 m × 500 mm

Looking for similar products? Visit Product Comparison Guide

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG
Parafilm is a registered trademark of Bemis Company, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phil M Novis et al.
Journal of phycology, 56(5), 1232-1244 (2020-05-13)
Lake snow, caused by the freshwater centric diatom Lindavia intermedia, has become problematic in several large, oligotrophic New Zealand lakes over the past decade. Macroaggregates produced by L. intermedia foul fishing lines, intake screens, and water filters, and have a negative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service