Key Documents
BR55579
BRAND® conical joint clip
PTFE, for NS 29/32
Synonym(s):
Conical joint clips, PTFE
About This Item
UNSPSC Code:
41100000
material
PTFE
description
Operating temperature up to 200 °C
with sealed-in steel spring
packaging
pack of 1 ea
manufacturer/tradename
BRAND 55579
General description
Will not scratch glass surfaces. Excellent resilience and stability. Outstanding chemical resistance.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)