Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BR43020

BRAND® glass bottle-top tilt pipette dispenser

volume 25 mL, with bottle

Synonym(s):

Automatic pipettes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

feature

Calibrated to deliver (TD, Ex)
with bottle

manufacturer/tradename

BRAND 43020

volume

25 mL

color

green markings

fitting

(29/32)

Looking for similar products? Visit Product Comparison Guide

General description

Calibrated to deliver (TD, Ex). Inscriptions in green enamel. The automatic dispenser is particularly well-suited for dispensing suspensions. Delivery range approximately +/- 10%.

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service