BR43016
BRAND® glass bottle-top tilt pipette dispenser
volume 20 mL, with bottle
Synonym(s):
Automatic pipettes
About This Item
feature
Calibrated to deliver (TD, Ex)
with bottle
manufacturer/tradename
BRAND 43016
volume
20 mL
color
green markings
fitting
(29/32)
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service