Key Documents
BR41010
BRAND® graduated beaker with handle and spout
polypropylene, graduation, embossed, 1000 mL, accuracy: 20 mL
Synonym(s):
Graduated Beakers, Plastics
About This Item
UNSPSC Code:
41121803
NACRES:
NB.43
material
polypropylene
feature
graduation embossed
manufacturer/tradename
BRAND 41010
measuring range
1000 mL, accuracy: 20 mL
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)