BR27843
BRAND® BLAUBRAND® ETERNA graduated pipette, calibrated to deliver
type 2, capacity 10 mL , with 0.1 ml subdivisions, suitable for cotton plugs
Synonym(s):
BLAUBRAND®, ETERNA, Graduated Pipettes, Pipettes
About This Item
material
AR-Glas®
agency
according to DIN 12 697
feature
class AS with batch number
graduation
packaging
pack of 12
manufacturer/tradename
BRAND 27843
measuring range
accuracy: 0.1 mL
capacity
10 mL , with 0.1 ml subdivisions
Looking for similar products? Visit Product Comparison Guide
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service