BR153020
BRAND® tubing connector, Y-shape (120°)
polypropylene, for tubing i.d., 10 ‑ 11 mm
Synonym(s):
Tubing, Tubing Connectors
About This Item
Recommended Products
material
polypropylene
feature
for tubing i.d. 10 ‑ 11 mm
packaging
pack of 20 ea
manufacturer/tradename
BRAND 153020
L
65 mm
Looking for similar products? Visit Product Comparison Guide
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service