BR144345
BRAND® stopper
silicone, H 30 mm, top diam. 32 mm
Synonym(s):
Stoppers
About This Item
material
silicone
packaging
pack of 10 ea
manufacturer/tradename
BRAND 144345
H
30 mm
bottom diam.
26 mm
top diam.
32 mm
Looking for similar products? Visit Product Comparison Guide
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service