Key Documents
BR130412
BRAND® square bottle with tamper-proof screw cap, PE-HD
capacity 100 mL, wide-neck
Synonym(s):
Bottles, Plastics, Square bottles
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
material
high-density polyethylene bottle
polypropylene cap
square bottle
translucent bottle
wide-mouth bottle
wide-neck
joint
Joints threaded neck (GL32)
feature
with cap
packaging
pack of 100 ea
manufacturer/tradename
BRAND 130412
L × H
45 mm × 101 mm
capacity
100 mL
General description
Translucent. Screw cap with sealing cone, PP, blue, with cadmium-free pigments. Tamper-proof closure with press-off retaining ring. Space-saving square base.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)