Key Documents
BR130262
BRAND® amber narrow-mouth bottle, LDPE, with screw cap
capacity 1000 mL
Synonym(s):
Bottles, Plastics
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
material
low-density polyethylene cap
round amber low-density polyethylene bottle
joint
Joints threaded neck (GL28)
feature
with cap
packaging
pack of 100 ea
manufacturer/tradename
BRAND 130262
capacity
1000 mL
diam. × H
94 mm × 212 mm
General description
For storage of light-sensitive substances. With screw cap
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)