BR110705
BRAND® mortar with pestle, with foot/spout
melamine formaldehyde, size 125 mm × 80 mm
Synonym(s):
Mortar
About This Item
Recommended Products
material
melamine formaldehyde
feature
autoclavable at 121°C
manufacturer/tradename
BRAND 110705
size
125 mm × 80 mm
Looking for similar products? Visit Product Comparison Guide
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service