Key Documents
BMSH3760HSE
Sigma-Aldrich
Digital Hotplate Magnetic Stirrer H3760
AC/DC input 230 V AC, Schuko plug
About This Item
UNSPSC Code:
41102406
NACRES:
NB.76
Product BMSH3760HSE is not currently sold in your country. Contact Technical Service
material
ceramic plate
description
Ceramic work surface, 6.5 x 6.5 in.
Large, backlit LCD display
manufacturer/tradename
Benchmark Scientific H3760-HS-E
parameter
200-1500 rpm speed
plate size × W × L
× 165 mm × 165 in.
General description
Benchmark hotplates, stirrers and hotplate-stirrers feature an exceptionally durable, chemical resistant, white ceramic work surface. Their space-efficient design (7 X 10.5 in. footprint), makes them ideal for use on crowded benchtops and inside bio-hoods.
Advanced microprocessor controls allow quick, precise adjustment of speed and temperature. The large, backlit LCD display provides constant visualization of the speed and temperature while a safety LED indicates when the top plate is above 50°C. In addition, an optional probe is available for applications that require precise temperature contol. When connected, the probe provides direct feedback to the microprocessor, which continuously adjusts and maintains the selected temperature in the sample within +/- 0.5°C.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)