Key Documents
BAH372500000
Micro-Mill grinder, blade and grinding chamber
stainless steel, AC/DC input 115 V AC
Synonym(s):
grinding mill, homoginizer mill
About This Item
UNSPSC Code:
41101708
material
stainless steel
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art H37250-0000
L × W × H
14 cm (5 1/2 in.) × 18 cm (7 in.) × 28 cm (11 in.)
weight
6.4 kg (14 lb)
General description
The grinders have a stainless steel or optional hard-faced blade rotating at up to 12,000 rpm to rapidly grind a wide variety of material including bauxite, bone, ceramics, coal, Devorda′s alloy and wood. Interchangeable grinding chambers reduce cross contamination.
Each chamber is 54 mm x 25 mm (2 1/8 in. x 1 in.) I.D. The 1/5 horsepower (150 Watts) motor develops a torque of 2.2 kg-cm (30 oz-inches) to maintain blade speed. The aluminum housing with epoxy finish is corrosion resistant and easy to clean. Built-in heat exchangers help maintain sample temperature. Included tubing connections, 6.4 mm (1/4 in.), on the top and housing can circulate water, methanol/dry ice slurry or liquid nitrogen for more aggressive sample cooling.
A resetable circuit breaker protects the motor and circuitry against damage from a jammed blade. Non-skid rubber feet protect the bench top. Power supplied through a 1.8 m (6 ft) 3-wire cord with grounded plug.
  • Rapid pulverization of small samples from 20cc to 50cc
  • Ideal for processing minerals, pharmaceuticals, pigment blends and other solid samples
  • Built in safety switching avoids inadvertent operation unless fully assembled
  • Cover, milling blade and grinding chamber are detachable for thorough cleaning
Legal Information
Micro-Mill is a trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)