Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BAH186610000

Lab Fridge tray rack

9 channels x 25 mm diam.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122806
NACRES:
NB.35

material

polypropylene body
polystyrene rack

feature

Holds 83 mm high bottles

packaging

pack of 1 ea

manufacturer/tradename

Bel-Art H18661-0000

technique(s)

cryopreservation: suitable

L × W × H

330 mm (13 in.) × 330 mm (13 in.) × 44 mm (1 3/4 in.)

Looking for similar products? Visit Product Comparison Guide

General description

Select from two channels widths to match vial size. Use in drawers to separate pipettes and stirring rods. vacuum formed white styrene. Organize storage of small bottles of blood-typing sera, chemistry controls, enzyme reagents and coagulant reagents.

  • Maximize refrigerator capacity

Legal Information

Lab Fridge is a trademark of SP Scienceware, a division of SP

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service